Product Details

SNP ID
rs369353928
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:248847238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGTCAGGAGGTGGACCCCAGAG[C/T]GTGAGTGGCACGCTTCCCTGTGAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF672 PubMed Links

Gene Details

Gene
ZNF672
Gene Name
zinc finger protein 672
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024836.2 812 UTR 5 NP_079112.1
XM_005270336.1 812 UTR 5 XP_005270393.1
Gene
ZNF692
Gene Name
zinc finger protein 692
There are no transcripts associated with this gene.

View Full Product Details