Product Details

SNP ID
rs8103270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9377785 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATATGCTCACCTTTATGGTCCTTA[C/T]GAGTTACTCCTTATTGTTCTAAGTA
Phenotype
MIM: 601276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF177 PubMed Links

Gene Details

Gene
ZNF177
Gene Name
zinc finger protein 177
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172651.1 Intron NP_001166122.1
NM_003451.2 Intron NP_003442.2
Gene
ZNF559-ZNF177
Gene Name
ZNF559-ZNF177 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172650.2 Intron NP_001166121.1
NM_001202425.1 Intron NP_001189354.1

View Full Product Details