Product Details

SNP ID
hCV27839141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102470633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCAGCCTGCCAAGTCTGACACG[G/T]CATCTCCCAACAACTCTCCTTTCCC
Phenotype
MIM: 605143
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACTR1A PubMed Links

Gene Details

Gene
ACTR1A
Gene Name
ARP1 actin-related protein 1 homolog A, centractin alpha
There are no transcripts associated with this gene.

Gene
MFSD13A
Gene Name
major facilitator superfamily domain containing 13A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024789.3 Intron NP_079065.2

View Full Product Details