Product Details

SNP ID
rs113979451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60060995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGGCAAGACAGAATTGAATATT[A/G]TACAAACTCTGTTACTCTGCCCTGG
Phenotype
MIM: 606498
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MS4A3 PubMed Links

Gene Details

Gene
MS4A3
Gene Name
membrane spanning 4-domains A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031666.1 Intron NP_001026836.1
NM_001031809.1 Intron NP_001026979.1
NM_006138.4 Intron NP_006129.4
XM_011545363.2 Intron XP_011543665.1

View Full Product Details