Product Details

SNP ID
rs60333007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:10694599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGCTGCCGGCGCTACAGCCCCTA[A/C]GCAACCGGCCGGAAGTCGGCCCCAC
Phenotype
MIM: 607811
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C6orf52 PubMed Links

Gene Details

Gene
C6orf52
Gene Name
chromosome 6 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145020.1 8 UTR 5 NP_001138492.1
XM_006715088.3 8 UTR 5 XP_006715151.1
XM_011514572.2 8 Intron XP_011512874.1
XM_011514574.2 8 Intron XP_011512876.1
XM_011514575.2 8 UTR 5 XP_011512877.1
Gene
PAK1IP1
Gene Name
PAK1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017906.2 8 Intron NP_060376.2
XM_005249204.2 8 Intron XP_005249261.1
XM_011514720.1 8 Intron XP_011513022.1
XM_011514721.1 8 Intron XP_011513023.1

View Full Product Details