Product Details

SNP ID
rs150784522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:5261101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGCCCGTGCATCCAGCCAAGCT[C/G]CTGGCGGACGGATATCGGAGGCCCA
Phenotype
MIM: 611592 MIM: 613311
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FARS2 PubMed Links

Gene Details

Gene
FARS2
Gene Name
phenylalanyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318872.1 101 UTR 5 NP_001305801.1
NM_006567.4 101 Intron NP_006558.1
XM_005248812.3 101 Intron XP_005248869.1
XM_006714966.2 101 Intron XP_006715029.1
XM_011514247.2 101 Intron XP_011512549.1
XM_011514248.2 101 Intron XP_011512550.1
XM_011514249.1 101 Intron XP_011512551.1
XM_011514251.2 101 Intron XP_011512553.1
XM_017010186.1 101 UTR 5 XP_016865675.1
XM_017010187.1 101 UTR 5 XP_016865676.1
Gene
LYRM4
Gene Name
LYR motif containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164840.2 101 Intron NP_001158312.1
NM_001164841.2 101 Intron NP_001158313.1
NM_001318782.1 101 Intron NP_001305711.1
NM_001318783.1 101 Intron NP_001305712.1
NM_020408.5 101 Intron NP_065141.3
XM_017011083.1 101 Intron XP_016866572.1
XM_017011084.1 101 Intron XP_016866573.1

View Full Product Details