Product Details

SNP ID
rs58913958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41689298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTTGGGCGGGCCCGGCGTCTC[A/G]GTGGCGCATTTACTAATGGCTCCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF1 PubMed Links

Gene Details

Gene
EIF1
Gene Name
eukaryotic translation initiation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005801.3 Intron NP_005792.1

View Full Product Details