Product Details
- SNP ID
-
rs112090903
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:227282 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGATTCAAGGCTTCCTCTCTGGCA[C/T]CCAACCATGAAGCTCCTTTTTCCTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB129
PubMed Links
Gene Details
- Gene
- DEFB129
- Gene Name
- defensin beta 129
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080831.3 |
25 |
UTR 5 |
|
|
NP_543021.1 |
View Full Product Details