Product Details

SNP ID
rs7024732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35658023 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAGGCCTTCAGCACGAACCACGT[C/T]CTCAGCTTCACAGAGTAGTATTTTA
Phenotype
MIM: 157660 MIM: 604964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF39 PubMed Links

Gene Details

Gene
ARHGEF39
Gene Name
Rho guanine nucleotide exchange factor 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032818.2 Intron NP_116207.2
XM_011518057.2 Intron XP_011516359.1
XM_017015225.1 Intron XP_016870714.1
XM_017015226.1 Intron XP_016870715.1
XM_017015227.1 Intron XP_016870716.1
XM_017015228.1 Intron XP_016870717.1
Gene
CCDC107
Gene Name
coiled-coil domain containing 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195200.1 Intron NP_001182129.1
NM_001195201.1 Intron NP_001182130.1
NM_001195217.1 Intron NP_001182146.1
NM_174923.2 Intron NP_777583.2
XM_005251403.4 Intron XP_005251460.1
Gene
RMRP
Gene Name
RNA component of mitochondrial RNA processing endoribonuclease
There are no transcripts associated with this gene.

Gene
SIT1
Gene Name
signaling threshold regulating transmembrane adaptor 1
There are no transcripts associated with this gene.

View Full Product Details