Product Details
- SNP ID
-
rs73959530
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:58863141 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCACGGTCTTTTCAACCATCTTTG[A/G]CGAGGCCTTGCTTCCTTCCACTCGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF532
PubMed Links
Gene Details
- Gene
- ZNF532
- Gene Name
- zinc finger protein 532
View Full Product Details