Product Details

SNP ID
rs188059204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47222030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCCTCCTTGCTGGCCCAGAGC[C/T]CAAGAGGAAGCAGAAGACGGCTGGC
Phenotype
MIM: 311550 MIM: 314370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK16 PubMed Links

Gene Details

Gene
CDK16
Gene Name
cyclin dependent kinase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170460.1 1122 Intron NP_001163931.1
NM_006201.4 1122 Intron NP_006192.1
NM_033018.3 1122 Intron NP_148978.2
XM_011543920.2 1122 Intron XP_011542222.1
XM_011543921.1 1122 Intron XP_011542223.1
XM_011543922.1 1122 Intron XP_011542224.1
XM_011543923.1 1122 Intron XP_011542225.1
XM_011543924.1 1122 Intron XP_011542226.1
XM_011543925.2 1122 Intron XP_011542227.1
XM_011543926.1 1122 UTR 5 XP_011542228.1
XM_011543927.2 1122 Intron XP_011542229.1
XM_011543928.1 1122 Intron XP_011542230.1
XM_017029569.1 1122 Intron XP_016885058.1
XM_017029570.1 1122 Intron XP_016885059.1
XM_017029571.1 1122 UTR 5 XP_016885060.1
XM_017029572.1 1122 Intron XP_016885061.1
XM_017029573.1 1122 Intron XP_016885062.1
Gene
UBA1
Gene Name
ubiquitin like modifier activating enzyme 1
There are no transcripts associated with this gene.

View Full Product Details