Product Details

SNP ID
rs148093305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:170358909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAAGAGAAAGTTAATTTTTGCC[C/T]GCAGTCATTCCAGAAAAATAGTGTA
Phenotype
MIM: 165340
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SKIL PubMed Links

Gene Details

Gene
SKIL
Gene Name
SKI-like proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145097.2 Intron NP_001138569.1
NM_001145098.2 Intron NP_001138570.1
NM_001248008.1 Intron NP_001234937.1
NM_005414.4 Intron NP_005405.2
XM_005247721.1 Intron XP_005247778.1
XM_006713735.1 Intron XP_006713798.1

View Full Product Details