Product Details

SNP ID
rs568659132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54200847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTACTATAAGAGCGCAGCCGTGG[C/G]GCTTCCGCGCCTCTTTCTCAGTGAC
Phenotype
MIM: 603631 MIM: 608754
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPS9 PubMed Links

Gene Details

Gene
RPS9
Gene Name
ribosomal protein S9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013.3 291 Intron NP_001004.2
NM_001321701.1 291 Intron NP_001308630.1
NM_001321702.1 291 Intron NP_001308631.1
NM_001321704.1 291 Intron NP_001308633.1
NM_001321705.1 291 Intron NP_001308634.1
NM_001321706.1 291 Intron NP_001308635.1
XM_017027112.1 291 UTR 5 XP_016882601.1
Gene
TSEN34
Gene Name
tRNA splicing endonuclease subunit 34
There are no transcripts associated with this gene.

View Full Product Details