Product Details

SNP ID
rs9942182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:127632830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGAGAAGCCCTCCTGGAAGTTT[A/C]AGCCCATTGGCTGCTTTCTGTTCTC
Phenotype
MIM: 610621
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
INTU PubMed Links

Gene Details

Gene
INTU
Gene Name
inturned planar cell polarity protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015693.3 Intron NP_056508.2
XM_011531844.2 Intron XP_011530146.1
XM_011531845.2 Intron XP_011530147.1
XM_011531846.1 Intron XP_011530148.1
XM_011531847.1 Intron XP_011530149.1
XM_011531848.2 Intron XP_011530150.1
XM_011531849.2 Intron XP_011530151.1
XM_011531850.2 Intron XP_011530152.1
XM_011531851.2 Intron XP_011530153.1
XM_017008025.1 Intron XP_016863514.1
XM_017008026.1 Intron XP_016863515.1

View Full Product Details