Product Details

SNP ID
rs77761746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:44067262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCCAGGCAATACTCTGCTTTAA[C/T]TCAGGAAATTTTAACCTGGCATTTG
Phenotype
MIM: 608405
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACCS PubMed Links

Gene Details

Gene
ACCS
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127219.1 553 Intron NP_001120691.1
NM_032592.3 553 Intron NP_115981.1
XM_005253170.3 553 Missense Mutation ACT,ATT T90I XP_005253227.2
XM_005253172.2 553 Intron XP_005253229.1
XM_006718348.2 553 Intron XP_006718411.1
XM_011520406.2 553 Missense Mutation ACT,ATT T90I XP_011518708.1
XM_011520407.1 553 Intron XP_011518709.1
XM_011520410.2 553 Intron XP_011518712.1
XM_017018421.1 553 Missense Mutation ACT,ATT T90I XP_016873910.1
Gene
ACCSL
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like
There are no transcripts associated with this gene.

View Full Product Details