Product Details

SNP ID
rs11569317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46122211 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCATGGAGTTGGCCAGAGCCCTCC[C/G]TCATTTCCTGATGTTTTCCAGGACA
Phenotype
MIM: 109535
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD40 PubMed Links

Gene Details

Gene
CD40
Gene Name
CD40 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001250.5 Intron NP_001241.1
NM_001302753.1 Intron NP_001289682.1
NM_001322421.1 Intron NP_001309350.1
NM_001322422.1 Intron NP_001309351.1
NM_152854.3 Intron NP_690593.1
XM_005260619.3 Intron XP_005260676.1
XM_011529109.2 Intron XP_011527411.1
XM_017028135.1 Intron XP_016883624.1
XM_017028136.1 Intron XP_016883625.1
XM_017028137.1 Intron XP_016883626.1

View Full Product Details