Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015310.3 | 10413 | UTR 3 | NP_056125.3 | ||
NM_206909.2 | 10413 | UTR 3 | NP_996792.1 | ||
XM_011544467.2 | 10413 | Intron | XP_011542769.1 | ||
XM_011544468.2 | 10413 | Intron | XP_011542770.1 | ||
XM_011544469.2 | 10413 | Intron | XP_011542771.1 | ||
XM_011544471.2 | 10413 | Intron | XP_011542773.1 | ||
XM_011544473.2 | 10413 | Intron | XP_011542775.1 | ||
XM_017013258.1 | 10413 | UTR 3 | XP_016868747.1 | ||
XM_017013259.1 | 10413 | UTR 3 | XP_016868748.1 | ||
XM_017013260.1 | 10413 | UTR 3 | XP_016868749.1 | ||
XM_017013261.1 | 10413 | UTR 3 | XP_016868750.1 | ||
XM_017013262.1 | 10413 | Intron | XP_016868751.1 | ||
XM_017013263.1 | 10413 | Intron | XP_016868752.1 | ||
XM_017013264.1 | 10413 | Intron | XP_016868753.1 | ||
XM_017013265.1 | 10413 | UTR 3 | XP_016868754.1 |