Product Details

SNP ID
rs76814569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35855158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACTCCTTGCCCTCCTCCCACCT[C/G]CCCTCTCCCAACTGCTCTCTACACC
Phenotype
MIM: 607762 MIM: 602716
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIRREL2 PubMed Links

Gene Details

Gene
KIRREL2
Gene Name
kin of IRRE like 2 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032123.6 Intron NP_115499.5
NM_199179.3 Intron NP_954648.3
NM_199180.3 Intron NP_954649.3
XM_011527362.1 Intron XP_011525664.1
XM_011527363.1 Intron XP_011525665.1
XM_011527365.1 Intron XP_011525667.1
XM_011527366.1 Intron XP_011525668.1
XM_017027348.1 Intron XP_016882837.1
Gene
NPHS1
Gene Name
NPHS1, nephrin
There are no transcripts associated with this gene.

View Full Product Details