Product Details

SNP ID
rs138696611
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:28553062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGGCGCCCGCGCGGGCCTTCGA[G/T]CCCCGCCCGCAGAGCCCCGCCCGCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRNAU1AP PubMed Links

Gene Details

Gene
TRNAU1AP
Gene Name
tRNA selenocysteine 1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017846.4 46 UTR 5 NP_060316.1
XM_017001542.1 46 Intron XP_016857031.1

View Full Product Details