Product Details

SNP ID
rs199916734
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.1:206507595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGCTGCCAAAGCTGCCGCCACT[A/G]GCCGGGCATGGCCATGGCGTCCCCG
Phenotype
MIM: 607020
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RASSF5 PubMed Links

Gene Details

Gene
RASSF5
Gene Name
Ras association domain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182663.3 66 UTR 5 NP_872604.1
NM_182664.3 66 UTR 5 NP_872605.1
NM_182665.3 66 Intron NP_872606.1

View Full Product Details