Product Details

SNP ID
rs181475306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:102147876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGGAACTAAAAAGAATCAAAA[A/T]TTTCCCCCATCAATAGTACTTAAGT
Phenotype
MIM: 613687
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PARPBP PubMed Links

Gene Details

Gene
PARPBP
Gene Name
PARP1 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319988.1 Intron NP_001306917.1
NM_001319993.1 Intron NP_001306922.1
NM_001319994.1 Intron NP_001306923.1
NM_001319995.1 Intron NP_001306924.1
NM_001319996.1 Intron NP_001306925.1
NM_017915.4 Intron NP_060385.3
XM_011538509.1 Intron XP_011536811.1
XM_011538514.2 Intron XP_011536816.1
XM_011538515.1 Intron XP_011536817.1
XM_017019534.1 Intron XP_016875023.1
XM_017019535.1 Intron XP_016875024.1
XM_017019536.1 Intron XP_016875025.1
XM_017019537.1 Intron XP_016875026.1
XM_017019538.1 Intron XP_016875027.1
XM_017019539.1 Intron XP_016875028.1
XM_017019540.1 Intron XP_016875029.1
XM_017019541.1 Intron XP_016875030.1
XM_017019542.1 Intron XP_016875031.1
XM_017019543.1 Intron XP_016875032.1
XM_017019544.1 Intron XP_016875033.1

View Full Product Details