Product Details

SNP ID
rs112194209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36486720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCATGGCGTCGCTCAAATGTAG[C/T]ACCGTCGTCTGCGTGATCTGCTTGG
Phenotype
MIM: 604500
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYO19 PubMed Links

Gene Details

Gene
MYO19
Gene Name
myosin XIX
There are no transcripts associated with this gene.

Gene
ZNHIT3
Gene Name
zinc finger HIT-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281432.1 94 Silent Mutation AGC,AGT S7S NP_001268361.1
NM_001281433.1 94 Silent Mutation AGC,AGT S7S NP_001268362.1
NM_001281434.1 94 Silent Mutation AGC,AGT S7S NP_001268363.1
NM_004773.3 94 Silent Mutation AGC,AGT S7S NP_004764.1

View Full Product Details