Product Details

SNP ID
rs191338538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:83853266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCACGCGCAGCCTTTTTGTTTAT[C/G]CGGCTAGCAGGGAGAGGGGCGGAGA
Phenotype
MIM: 608343 MIM: 609891
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYB5R4 PubMed Links

Gene Details

Gene
CYB5R4
Gene Name
cytochrome b5 reductase 4
There are no transcripts associated with this gene.

Gene
RIPPLY2
Gene Name
ripply transcriptional repressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009994.2 1 UTR 5 NP_001009994.1

View Full Product Details