Product Details

SNP ID
rs73944350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:102473426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATGCAGTGAAATGAGGAAATGC[C/T]TTTAAGTGAAGAGATTTTTATTTCT
Phenotype
MIM: 600531
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC9A4 PubMed Links

Gene Details

Gene
SLC9A4
Gene Name
solute carrier family 9 member A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011552.3 145 UTR 5 NP_001011552.2
XM_011511158.1 145 UTR 5 XP_011509460.1

View Full Product Details