Product Details

SNP ID
rs867115209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31848985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAACCCACTGGTGCCCATCTTG[C/T]GCTCATTTCCCATGCCCAAGTCCTG
Phenotype
MIM: 616776 MIM: 602939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP15 PubMed Links

Gene Details

Gene
DUSP15
Gene Name
dual specificity phosphatase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012644.2 Intron NP_001012662.1
NM_001320478.1 Intron NP_001307407.1
NM_001320479.1 Intron NP_001307408.1
NM_080611.4 Intron NP_542178.2
NM_177991.2 Intron NP_817130.1
XM_017027654.1 Intron XP_016883143.1
XM_017027655.1 Intron XP_016883144.1
XM_017027656.1 Intron XP_016883145.1
XM_017027657.1 Intron XP_016883146.1
XM_017027658.1 Intron XP_016883147.1
XM_017027659.1 Intron XP_016883148.1
XM_017027660.1 Intron XP_016883149.1
XM_017027661.1 Intron XP_016883150.1
XM_017027662.1 Intron XP_016883151.1
Gene
FOXS1
Gene Name
forkhead box S1
There are no transcripts associated with this gene.

View Full Product Details