Product Details

SNP ID
rs7986690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49630195 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAGACTTTTCCTGTTTTGGAAAA[A/G]CTCTTCCGTTGTTTGCCTGCACAGA
Phenotype
MIM: 609351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL11 PubMed Links

Gene Details

Gene
ARL11
Gene Name
ADP ribosylation factor like GTPase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138450.5 Intron NP_612459.1
XM_005266253.3 Intron XP_005266310.1

View Full Product Details