Product Details

SNP ID
rs9931575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10529253 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGTCCTAATATGTGCTTCTTGGA[A/T]GTCTACTAATTTTGTTGTTGTTATT
Phenotype
MIM: 602334
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EMP2 PubMed Links

Gene Details

Gene
EMP2
Gene Name
epithelial membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001424.5 4378 UTR 3 NP_001415.1
XM_006720864.3 4378 UTR 3 XP_006720927.1

View Full Product Details