Product Details

SNP ID
rs121434262
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:193122225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCGGACGTGCTTAGCGTCCTG[C/T]GACAGTACAACATCCAGAAGAAGGA
Phenotype
MIM: 607393
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC73 PubMed Links

Gene Details

Gene
CDC73
Gene Name
cell division cycle 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024529.4 268 Nonsense Mutation CGA,TGA R9* NP_078805.3
XM_006711537.3 268 Nonsense Mutation CGA,TGA R9* XP_006711600.1

View Full Product Details