Product Details

SNP ID
hCV27860237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:161306404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGTAGAAAAGCAGCAGCAGCAAC[A/C]GCACCACCCCGAGGACACCCCCGAT
Phenotype
MIM: 159440 MIM: 602413
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MPZ PubMed Links

Gene Details

Gene
MPZ
Gene Name
myelin protein zero
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000530.7 576 Missense Mutation CGG,CTG R170L NP_000521.2
NM_001315491.1 576 Missense Mutation CGG,CTG R170L NP_001302420.1
Gene
SDHC
Gene Name
succinate dehydrogenase complex subunit C
There are no transcripts associated with this gene.

View Full Product Details