Product Details
- SNP ID
-
hCV27862819
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44374661 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCTCCATCCCCCCACACTCACCT[A/G]AGCTTCCCCTCGGGGCAGGCTGAGC
- Phenotype
-
MIM: 607759
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ITGA2B
PubMed Links
Gene Details
- Gene
- ITGA2B
- Gene Name
- integrin subunit alpha 2b
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000419.4 |
3069 |
Nonsense Mutation |
CAG,TAG |
Q981* |
NP_000410.2 |
View Full Product Details