Product Details

SNP ID
rs104894273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112226517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCATCTCCTTCAGCGCGAGCCACC[A/G]ATTGTACAGGTAGGGTGTGCACACA
Phenotype
MIM: 611740 MIM: 612719
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCO2 PubMed Links

Gene Details

Gene
BCO2
Gene Name
beta-carotene oxygenase 2
There are no transcripts associated with this gene.

Gene
PTS
Gene Name
6-pyruvoyltetrahydropterin synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000317.2 153 Missense Mutation CAA,CGA Q25R NP_000308.1

View Full Product Details