Product Details

SNP ID
rs4549258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:14952335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTGAGCCACTGGTCTGTGTCTA[A/G]CTGCCTGAGTGTCTCCACATATCGA
Phenotype
MIM: 601426
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGD5-AS1 PubMed Links

Gene Details

Gene
FGD5-AS1
Gene Name
FGD5 antisense RNA 1
There are no transcripts associated with this gene.

Gene
NR2C2
Gene Name
nuclear receptor subfamily 2 group C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291694.1 Intron NP_001278623.1
NM_003298.4 Intron NP_003289.2

View Full Product Details