Product Details
- SNP ID
-
rs4459515
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:78091675 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGCCCTAACTGCTCCAAAGCTTC[C/T]TACCACAGGCCTATCCCTGCCTTTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SH2D7
PubMed Links
Gene Details
- Gene
- SH2D7
- Gene Name
- SH2 domain containing 7
View Full Product Details