Product Details
- SNP ID
-
rs4893635
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:52631981 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAGGCCCAAGAAAAGGAAAAGAGT[T/C]GGGAAGCGCTCATCGGTGGAGCAGT
- Phenotype
-
MIM: 300543
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SSX8
PubMed Links
Gene Details
- Gene
- SSX8
- Gene Name
- SSX family member 8
There are no transcripts associated with this gene.
View Full Product Details