Product Details

SNP ID
rs4681676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58314070 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCATGTCTCAAAAATAATAACAC[A/G]TGGGGCGTGGTGGCTCATGCCTGTA
Phenotype
MIM: 606112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPP14 PubMed Links

Gene Details

Gene
RPP14
Gene Name
ribonuclease P/MRP subunit p14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098783.2 Intron NP_001092253.1
NM_007042.4 Intron NP_008973.1
XM_011533313.2 Intron XP_011531615.1
XM_017005641.1 Intron XP_016861130.1

View Full Product Details