Product Details

SNP ID
rs4415284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:379000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTTCCTTCCCTGTCCCAGCCC[C/T]ACAGACACATAACTGAGAAGGGGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM87A PubMed Links

Gene Details

Gene
FAM87A
Gene Name
family with sequence similarity 87 member A
There are no transcripts associated with this gene.

View Full Product Details