Product Details

SNP ID
rs4727325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:95774651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTATATCTCAGAGAAGGCTCTA[C/G]GGTCTTTCTTCCCAATCCCTCTGCT
Phenotype
MIM: 603772
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DYNC1I1 PubMed Links

Gene Details

Gene
DYNC1I1
Gene Name
dynein cytoplasmic 1 intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135556.1 Intron NP_001129028.1
NM_001135557.1 Intron NP_001129029.1
NM_001278421.1 Intron NP_001265350.1
NM_001278422.1 Intron NP_001265351.1
NM_004411.4 Intron NP_004402.1
XM_011515861.1 Intron XP_011514163.1
XM_011515862.1 Intron XP_011514164.1
XM_011515863.2 Intron XP_011514165.1
XM_017011804.1 Intron XP_016867293.1
XM_017011805.1 Intron XP_016867294.1
XM_017011806.1 Intron XP_016867295.1
XM_017011807.1 Intron XP_016867296.1

View Full Product Details