Product Details
- SNP ID
-
rs4895523
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:138426481 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTGACTGTGTATTTTTCCTATCAC[A/C]AAAAGGGGAATAACAGTATATAGGT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3145
PubMed Links
Gene Details
- Gene
- MIR3145
- Gene Name
- microRNA 3145
There are no transcripts associated with this gene.
- Gene
- NHSL1
- Gene Name
- NHS like 1
View Full Product Details