Product Details
- SNP ID
-
rs4920579
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
29
- Location
-
Chr.1:17251870 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCTGCCATATGCAGGATGTCTGGG[A/G]AACAGTCTCTGGACCGTATTGGGAG
- Phenotype
-
MIM: 607934
MIM: 606755
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PADI1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs191059951] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PADI1
- Gene Name
- peptidyl arginine deiminase 1
There are no transcripts associated with this gene.
- Gene
- PADI3
- Gene Name
- peptidyl arginine deiminase 3
View Full Product Details