Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001044305.2 | 81 | Intron | NP_001037770.1 | ||
NM_001281439.1 | 81 | Intron | NP_001268368.1 | ||
NM_001281440.1 | 81 | UTR 5 | NP_001268369.1 | ||
NM_021940.4 | 81 | Intron | NP_068759.2 | ||
XM_005248760.4 | 81 | Intron | XP_005248817.1 | ||
XM_011536045.2 | 81 | Intron | XP_011534347.1 | ||
XM_011536046.2 | 81 | Intron | XP_011534348.1 | ||
XM_011536047.2 | 81 | Intron | XP_011534349.1 | ||
XM_011536048.2 | 81 | Intron | XP_011534350.1 | ||
XM_017011170.1 | 81 | Intron | XP_016866659.1 |