Product Details

SNP ID
rs4820671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20920862 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTATAGGGCCTTTCTCTGACCCT[A/C]CCCCTCACGTTCAGCTTTGTTTCTC
Phenotype
MIM: 602007
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRKL PubMed Links

Gene Details

Gene
CRKL
Gene Name
CRK like proto-oncogene, adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005207.3 Intron NP_005198.1

View Full Product Details