Product Details
- SNP ID
-
rs4839131
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
51
- Location
-
Chr.1:111389219 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGGATAGTCCATAGGTTTGTCCAT[C/T]GTTTCTGAAGGTGGAGGACAGGCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PGCP1
PubMed Links
Gene Details
- Gene
- PGCP1
- Gene Name
- progastricsin pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details