Product Details

SNP ID
rs4872423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:26299984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAACACTAGCATTACTACTGAAGT[C/G]TAGTAGACCTCACTCAGTACAGTCA
Phenotype
MIM: 604941
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPP2R2A PubMed Links

Gene Details

Gene
PPP2R2A
Gene Name
protein phosphatase 2 regulatory subunit Balpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177591.1 Intron NP_001171062.1
NM_002717.3 Intron NP_002708.1

View Full Product Details