Product Details
- SNP ID
-
rs4981084
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:30898855 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTTAAGCCCTTTTACTGGCTAACA[C/T]TGTGGGCCTCTATTAAATGGAGAGA
- Phenotype
-
MIM: 603196
MIM: 614766
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COCH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10047928] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COCH
- Gene Name
- cochlin
There are no transcripts associated with this gene.
- Gene
- LOC100506071
- Gene Name
- uncharacterized LOC100506071
There are no transcripts associated with this gene.
- Gene
- STRN3
- Gene Name
- striatin 3
View Full Product Details