Product Details

SNP ID
rs4830990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15742996 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCAACAGTGCCAAGACTGAGAAA[C/T]CCTAACTTAGAGAAAGAACCCTTGC
Phenotype
MIM: 300230
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CA5B PubMed Links

Gene Details

Gene
CA5B
Gene Name
carbonic anhydrase 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007220.3 Intron NP_009151.1
XM_005274442.4 Intron XP_005274499.1
XM_017029247.1 Intron XP_016884736.1
XM_017029248.1 Intron XP_016884737.1
XM_017029249.1 Intron XP_016884738.1

View Full Product Details