Product Details

SNP ID
rs4351535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107713231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCAGACACCAAGTTTATTTTTA[C/G]AAAAACAGAGCAAAGTTTATTGAAA
Phenotype
MIM: 300506
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TSC22D3 PubMed Links

Gene Details

Gene
TSC22D3
Gene Name
TSC22 domain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015881.1 2294 UTR 3 NP_001015881.1
NM_001318468.1 2294 UTR 3 NP_001305397.1
NM_001318470.1 2294 UTR 3 NP_001305399.1
NM_004089.3 2294 UTR 3 NP_004080.2
NM_198057.2 2294 UTR 3 NP_932174.1
XM_005262099.1 2294 UTR 3 XP_005262156.1
XM_005262100.1 2294 UTR 3 XP_005262157.1
XM_005262102.1 2294 UTR 3 XP_005262159.1
XM_005262103.3 2294 UTR 3 XP_005262160.1
XM_011530884.1 2294 UTR 3 XP_011529186.1
XM_017029335.1 2294 UTR 3 XP_016884824.1

View Full Product Details