Product Details

SNP ID
rs4711350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33773939 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGGTAAGTCAAGAGAGCAAACAG[A/G]GCATGGGCGTCTGGGGTTCACATGG
Phenotype
MIM: 616312
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LEMD2 PubMed Links

Gene Details

Gene
LEMD2
Gene Name
LEM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143944.1 Intron NP_001137416.1
NM_181336.3 Intron NP_851853.1
XM_017010437.1 Intron XP_016865926.1

View Full Product Details