Product Details

SNP ID
rs4821815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38709702 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCGTGATTCTTCTTGGTGTCAGCA[A/G]AGAAAAGATGCCCAGAATTCCTCTC
Phenotype
MIM: 602245 MIM: 615323
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTPBP1 PubMed Links

Gene Details

Gene
GTPBP1
Gene Name
GTP binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004286.4 Intron NP_004277.2
XM_011530537.2 Intron XP_011528839.1
XM_017029099.1 Intron XP_016884588.1
XM_017029100.1 Intron XP_016884589.1
XM_017029101.1 Intron XP_016884590.1
XM_017029102.1 Intron XP_016884591.1
XM_017029103.1 Intron XP_016884592.1
Gene
JOSD1
Gene Name
Josephin domain containing 1
There are no transcripts associated with this gene.

View Full Product Details