Product Details

SNP ID
rs4300409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74015503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAAGTCCCATTCCTGACCCCCAG[C/G]TGTACTGCAGGTAGATTTTCTGGGT
Phenotype
MIM: 615944 MIM: 602044
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C2CD3 PubMed Links

Gene Details

Gene
C2CD3
Gene Name
C2 calcium dependent domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286577.1 Intron NP_001273506.1
NM_015531.5 Intron NP_056346.3
Gene
UCP3
Gene Name
uncoupling protein 3
There are no transcripts associated with this gene.

View Full Product Details