Product Details

SNP ID
rs4793082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44187594 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTTCCTGTAAATCATGGTTAAT[A/G]ACATTAACCTTCTTACCATCAGGGG
Phenotype
MIM: 615056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB16 PubMed Links

Gene Details

Gene
ASB16
Gene Name
ankyrin repeat and SOCS box containing 16
There are no transcripts associated with this gene.

Gene
ASB16-AS1
Gene Name
ASB16 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ATXN7L3
Gene Name
ataxin 7 like 3
There are no transcripts associated with this gene.

Gene
TMUB2
Gene Name
transmembrane and ubiquitin like domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076674.1 389 Intron NP_001070142.1
NM_024107.2 389 Intron NP_077012.2
NM_177441.2 389 UTR 5 NP_803190.2
XM_011525198.2 389 UTR 5 XP_011523500.1
XM_011525200.2 389 Intron XP_011523502.1
XM_011525201.2 389 Intron XP_011523503.1
XM_011525203.2 389 Intron XP_011523505.1
XM_011525204.2 389 Intron XP_011523506.1
XM_011525205.2 389 UTR 5 XP_011523507.1
XM_011525206.2 389 Intron XP_011523508.1
XM_011525207.2 389 Intron XP_011523509.1
XM_011525208.2 389 Intron XP_011523510.1
XM_011525209.2 389 Intron XP_011523511.1
XM_011525210.2 389 Intron XP_011523512.1
XM_011525211.2 389 Intron XP_011523513.1
XM_017025047.1 389 UTR 5 XP_016880536.1
XM_017025048.1 389 Intron XP_016880537.1
XM_017025049.1 389 UTR 5 XP_016880538.1
XM_017025050.1 389 Intron XP_016880539.1
XM_017025051.1 389 Intron XP_016880540.1
XM_017025052.1 389 UTR 5 XP_016880541.1
XM_017025053.1 389 Intron XP_016880542.1
XM_017025054.1 389 UTR 5 XP_016880543.1
XM_017025055.1 389 Intron XP_016880544.1
XM_017025056.1 389 Intron XP_016880545.1

View Full Product Details